Assay Details
| Gene Symbol: Nt5c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164363.1 | NP_001157835.1 | 12 - 13 | 1428 | 88 | |
| NM_001164365.1 | NP_001157837.1 | 12 - 13 | 1085 | 88 | ||
| NM_029810.4 | NP_084086.3 | 12 - 13 | 1016 | 88 | ||
| NR_028353.1 | 13 - 14 | 1075 | 88 | |||
| GenBank mRNA | AK008045.1 | - | 12 - 13 | 963 | 88 | |
| AK030786.1 | - | 13 - 14 | 1035 | 88 | ||
| AK036730.1 | - | 12 - 13 | 974 | 88 | ||
| AK038458.1 | - | 9 - 10 | 722 | 88 | ||
| AK132384.1 | - | 12 - 13 | 974 | 88 | ||
| AK147901.1 | - | 12 - 13 | 974 | 88 | ||
| AK162385.1 | - | 12 - 13 | 1393 | 88 | ||
| AK169616.1 | - | 12 - 13 | 974 | 88 | ||
| AK193702.1 | - | 1 - 2 | 90 | 89 | ||
| AK204349.1 | - | 2 - 3 | 155 | 88 | ||
| BC064760.1 | - | 11 - 12 | 950 | 30436992 | 88 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BC006028.1 | - | Chr. - - [-] on Build | 88 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c2
Genomic Map