Assay Details
| Gene Symbol: Eif4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033561.1 | NP_291039.1 | 2 - 3 | 272 | 76 | |
| GenBank mRNA | AK045004.1 | - | 2 - 3 | 274 | 76 | |
| AK088818.1 | - | 2 - 3 | 284 | 76 | ||
| AK122194.1 | - | 2 - 3 | 272 | 76 | ||
| AK132034.1 | - | 2 - 3 | 292 | 76 | ||
| AK135390.1 | - | 2 - 3 | 273 | 76 | ||
| AK136999.1 | - | 2 - 3 | 259 | 76 | ||
| AK149841.1 | - | 2 - 3 | 258 | 76 | ||
| AK150088.1 | - | 2 - 3 | 281 | 76 | ||
| AK166567.1 | - | 2 - 3 | 273 | 76 | ||
| AK168872.1 | - | 2 - 3 | 291 | 76 | ||
| AK169312.1 | - | 2 - 3 | 255 | 76 | ||
| AK169353.1 | - | 2 - 3 | 273 | 76 | ||
| AK169407.1 | - | 2 - 3 | 291 | 76 | ||
| AK177188.1 | - | 1 - 2 | 33 | 76 | ||
| AK185103.1 | - | 1 - 2 | 76 | |||
| AK185835.1 | - | 1 - 2 | 109 | 76 | ||
| BC014796.1 | - | 2 - 3 | 286 | 3962104 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif4h
Genomic Map