Assay Details
| Gene Symbol: Fbxw7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177773.1 | NP_001171244.1 | 8 - 9 | 1368 | 118 | |
| NM_001177774.1 | NP_001171245.1 | 7 - 8 | 1277 | 118 | ||
| NM_080428.3 | NP_536353.2 | 5 - 6 | 913 | 118 | ||
| GenBank mRNA | AB221593.1 | - | 5 - 6 | 755 | 118 | |
| AF391192.1 | - | 5 - 6 | 913 | 118 | ||
| AF427101.1 | - | 5 - 6 | 835 | 118 | ||
| BC060620.1 | - | 8 - 9 | 1368 | 118 | ||
| BC066093.1 | - | 7 - 8 | 1277 | 118 | ||
| BC131648.1 | - | 5 - 6 | 777 | 40110349 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxw7
Genomic Map