Assay Details
| Gene Symbol: Fbxw7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177773.1 | NP_001171244.1 | 11 - 12 | 1802 | 68 | |
| NM_001177774.1 | NP_001171245.1 | 10 - 11 | 1711 | 68 | ||
| NM_080428.3 | NP_536353.2 | 8 - 9 | 1347 | 68 | ||
| GenBank mRNA | AB221593.1 | - | 8 - 9 | 1189 | 68 | |
| AF391192.1 | - | 8 - 9 | 1347 | 68 | ||
| AF427101.1 | - | 8 - 9 | 1269 | 68 | ||
| AK032174.1 | - | 2 - 3 | 273 | 68 | ||
| BC060620.1 | - | 11 - 12 | 1802 | 68 | ||
| BC066093.1 | - | 10 - 11 | 69 | |||
| BC085184.1 | - | 10 - 11 | 3503 | 30664813 | 68 | |
| BC131648.1 | - | 8 - 9 | 1211 | 40110349 | 68 | |
More Information
Set Membership: |
|

Fbxw7
Genomic Map