Assay Details
| Gene Symbol: Glmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161738.1 | NP_001155210.1 | 10 - 11 | 1122 | 147 | |
| NM_001161739.1 | NP_001155211.1 | 10 - 11 | 1136 | 147 | ||
| NM_133248.2 | NP_573511.2 | 10 - 11 | 1136 | 147 | ||
| GenBank mRNA | AJ566083.1 | - | 9 - 10 | 1004 | 147 | |
| AK034160.1 | - | 10 - 11 | 1123 | 147 | ||
| AK165661.1 | - | 10 - 11 | 1107 | 147 | ||
| AK169045.1 | - | 10 - 11 | 1108 | 147 | ||
| AK172659.1 | - | 10 - 11 | 1074 | 147 | ||
| BC003446.1 | - | 10 - 11 | 1072 | 2651614 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glmn
Genomic Map