Assay Details
| Gene Symbol: Sptssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164210.2 | NP_001157682.1 | 3 - 4 | 194 | 69 | |
| NM_001286959.1 | NP_001273888.1 | 2 - 3 | 163 | 69 | ||
| NM_133675.3 | NP_598436.1 | 2 - 3 | 163 | 69 | ||
| GenBank mRNA | AF365876.1 | - | 2 - 3 | 165 | 69 | |
| AK004017.1 | - | 2 - 3 | 165 | 69 | ||
| AK009973.1 | - | 3 - 4 | 161 | 69 | ||
| AK010117.1 | - | 2 - 3 | 164 | 69 | ||
| BC022628.1 | - | 2 - 3 | 143 | 4206004 | 69 | |
| CX729774.1 | - | 2 - 3 | 137 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF365876.1, AK004017.1, AK009973.1, AK010117.1, BC022628.1, NM_001164210.2, NM_001286959.1, NM_133675.3 transcripts.
Set Membership: |
|

Sptssb
Genomic Map