Assay Details
| Gene Symbol: Tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133683.3 | NP_598444.1 | 1 - 2 | 211 | 73 | |
| GenBank mRNA | AK034298.1 | - | 1 - 2 | 217 | 73 | |
| AK138931.1 | - | 1 - 2 | 199 | 73 | ||
| AK155275.1 | - | 1 - 2 | 172 | 73 | ||
| AK170143.1 | - | 1 - 2 | 171 | 73 | ||
| AK171087.1 | - | 1 - 2 | 171 | 73 | ||
| BC016895.1 | - | 1 - 2 | 213 | 4239007 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK034298.1,AK138931.1,AK155275.1,AK170143.1,AK171087.1,BC016895.1,NM_133683.3 transcripts.
Set Membership: |
|

Tmem19
Genomic Map