Assay Details
| Gene Symbol: Puf60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164600.1 | NP_001158072.1 | 7 - 8 | 820 | 77 | |
| NM_028364.2 | NP_082640.2 | 8 - 9 | 901 | 77 | ||
| NM_133691.5 | NP_598452.2 | 7 - 8 | 850 | 77 | ||
| GenBank mRNA | AK012434.1 | - | 7 - 8 | 790 | 77 | |
| AK075960.1 | - | 7 - 8 | 818 | 77 | ||
| AK146534.1 | - | 8 - 9 | 872 | 77 | ||
| AK149632.1 | - | 8 - 9 | 867 | 77 | ||
| AK151526.1 | - | 7 - 8 | 814 | 77 | ||
| AK168789.1 | - | 7 - 8 | 817 | 77 | ||
| BC010601.1 | - | 7 - 8 | 820 | 3257153 | 77 | |
| BC119437.1 | - | 5 - 6 | 557 | 77 | ||
| BC148274.1 | - | 5 - 6 | 557 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Puf60
Genomic Map