Assay Details
| Gene Symbol: Atp6v1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159632.1 | NP_001153104.1 | 9 - 10 | 809 | 83 | |
| NM_133699.2 | NP_598460.1 | 8 - 9 | 779 | 83 | ||
| GenBank mRNA | AB088357.1 | - | 8 - 9 | 764 | 83 | |
| AK004157.1 | - | 5 - 6 | 494 | 83 | ||
| BC056636.1 | - | 9 - 10 | 782 | 5065583 | 83 | |
| BC064071.1 | - | 8 - 9 | 748 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1c2
Genomic Map