Assay Details
| Gene Symbol: Atp6v1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159632.1 | NP_001153104.1 | 11 - 12 | 1000 | 77 | |
| NM_133699.2 | NP_598460.1 | 10 - 11 | 970 | 77 | ||
| GenBank mRNA | AB088357.1 | - | 10 - 11 | 955 | 77 | |
| AK004157.1 | - | 7 - 8 | 685 | 77 | ||
| BC056636.1 | - | 11 - 12 | 973 | 5065583 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1c2
Genomic Map