Assay Details
| Gene Symbol: Dcaf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199009.1 | NP_001185938.1 | 4 - 5 | 670 | 100 | |
| NM_133734.3 | NP_598495.1 | 4 - 5 | 575 | 100 | ||
| NR_037572.1 | 4 - 5 | 687 | 100 | |||
| GenBank mRNA | AK033540.1 | - | 4 - 5 | 573 | 100 | |
| AK152395.1 | - | 3 - 4 | 628 | 100 | ||
| AK158784.1 | - | 4 - 5 | 655 | 100 | ||
| AK170657.1 | - | 4 - 5 | 573 | 100 | ||
| BC008545.1 | - | 3 - 4 | 468 | 3497641 | 100 | |
| BC021305.1 | - | 2 - 3 | 2167 | 100 | ||
| BC037001.1 | - | 3 - 4 | 599 | 5051353 | 100 | |
| DV648268.1 | - | 4 - 5 | 670 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf11
Genomic Map