Assay Details
| Gene Symbol: Dcaf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199009.1 | NP_001185938.1 | 6 - 7 | 841 | 75 | |
| NM_133734.3 | NP_598495.1 | 6 - 7 | 746 | 75 | ||
| NR_037572.1 | 6 - 7 | 858 | 75 | |||
| GenBank mRNA | AK033540.1 | - | 6 - 7 | 744 | 75 | |
| AK150183.1 | - | 6 - 7 | 833 | 75 | ||
| AK152395.1 | - | 5 - 6 | 799 | 75 | ||
| AK158784.1 | - | 6 - 7 | 826 | 75 | ||
| AK170657.1 | - | 6 - 7 | 744 | 75 | ||
| BC008545.1 | - | 5 - 6 | 639 | 3497641 | 75 | |
| BC021305.1 | - | 4 - 5 | 2338 | 75 | ||
| BC037001.1 | - | 5 - 6 | 770 | 5051353 | 75 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK003028.1 | - | Chr. - - [-] on Build | 75 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf11
Genomic Map