Assay Details
| Gene Symbol: Dcaf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199009.1 | NP_001185938.1 | 7 - 8 | 983 | 74 | |
| NM_133734.3 | NP_598495.1 | 7 - 8 | 888 | 74 | ||
| NR_037572.1 | 7 - 8 | 1000 | 74 | |||
| GenBank mRNA | AK033540.1 | - | 7 - 8 | 886 | 74 | |
| AK150183.1 | - | 7 - 8 | 975 | 74 | ||
| AK152395.1 | - | 6 - 7 | 941 | 74 | ||
| AK158784.1 | - | 7 - 8 | 968 | 74 | ||
| BC008545.1 | - | 6 - 7 | 781 | 3497641 | 74 | |
| BC021305.1 | - | 5 - 6 | 2480 | 74 | ||
| BC037001.1 | - | 6 - 7 | 912 | 5051353 | 74 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK003028.1 | - | Chr. - - [-] on Build | 74 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf11
Genomic Map