Assay Details
| Gene Symbol: Fblim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163256.1 | NP_001156728.1 | 5 - 6 | 684 | 64 | |
| NM_133754.5 | NP_598515.3 | 5 - 6 | 739 | 64 | ||
| GenBank mRNA | AK010660.1 | - | 5 - 6 | 748 | 64 | |
| AK154680.1 | - | 5 - 6 | 720 | 64 | ||
| AK163587.1 | - | 5 - 6 | 725 | 64 | ||
| AK170151.1 | - | 5 - 6 | 739 | 64 | ||
| AK170440.1 | - | 5 - 6 | 689 | 64 | ||
| BC004777.1 | - | 4 - 5 | 614 | 3257486 | 64 | |
| CX563302.1 | - | 5 - 6 | 641 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fblim1
Genomic Map