Assay Details
| Gene Symbol: Mtif2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282118.1 | NP_001269047.1 | 9 - 10 | 1488 | 96 | |
| NM_001282119.1 | NP_001269048.1 | 9 - 10 | 1410 | 96 | ||
| NM_001282120.1 | NP_001269049.1 | 9 - 10 | 1852 | 96 | ||
| NM_133767.3 | NP_598528.2 | 9 - 10 | 1766 | 96 | ||
| GenBank mRNA | AK136344.1 | - | 9 - 10 | 1450 | 96 | |
| AK146961.1 | - | 9 - 10 | 1706 | 96 | ||
| BC016590.1 | - | 9 - 10 | 1342 | 3585359 | 96 | |
| BC066819.1 | - | 9 - 10 | 1746 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtif2
Genomic Map