Assay Details
| Gene Symbol: Gtf2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133801.2 | NP_598562.1 | 7 - 8 | 1006 | 64 | |
| GenBank mRNA | AK168236.1 | - | 7 - 8 | 1019 | 64 | |
| AK168243.1 | - | 7 - 8 | 1019 | 64 | ||
| BC003336.1 | - | 7 - 8 | 1003 | 64 | ||
| BC023081.1 | - | 7 - 8 | 1006 | 5355475 | 64 | |
| BC031123.1 | - | 7 - 8 | 1136 | 5026448 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2f1
Genomic Map