Assay Details
| Gene Symbol: Gtf2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133801.2 | NP_598562.1 | 10 - 11 | 1265 | 62 | |
| GenBank mRNA | AK168236.1 | - | 10 - 11 | 1278 | 62 | |
| AK168243.1 | - | 10 - 11 | 1278 | 62 | ||
| BC003336.1 | - | 10 - 11 | 1262 | 62 | ||
| BC023081.1 | - | 10 - 11 | 1265 | 5355475 | 62 | |
| BC031123.1 | - | 10 - 11 | 1395 | 5026448 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2f1
Genomic Map