Assay Details
| Gene Symbol: Uap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036243.1 | - | 8 - 9 | 1549 | 103 | |
| AK147139.1 | - | 8 - 9 | 1549 | 103 | ||
| AK171925.1 | - | 8 - 9 | 1446 | 103 | ||
| BC016406.1 | - | 8 - 9 | 1536 | 4489112 | 103 | |
More Information
Set Membership: |
|

Uap1
Genomic Map