Assay Details
| Gene Symbol: Kmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133809.1 | NP_598570.1 | 1 - 2 | 86 | 99 | |
| GenBank mRNA | AK129011.1 | - | 99 | 99 | ||
| AK158251.1 | - | 1 - 2 | 281 | 99 | ||
| AK171812.1 | - | 1 - 2 | 210 | 99 | ||
| BC014683.1 | - | 1 - 2 | 86 | 4210278 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kmo
Genomic Map