Assay Details
| Gene Symbol: Kmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133809.1 | NP_598570.1 | 8 - 9 | 717 | 102 | |
| GenBank mRNA | AK158251.1 | - | 8 - 9 | 913 | 102 | |
| BC014683.1 | - | 8 - 9 | 717 | 4210278 | 102 | |
More Information
Set Membership: |
|

Kmo
Genomic Map