Assay Details
| Gene Symbol: Ifi44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133871.2 | NP_598632.2 | 5 - 6 | 1005 | 72 | |
| GenBank mRNA | AJ299405.1 | - | 4 - 5 | 895 | 72 | |
| AK079749.1 | - | 5 - 6 | 957 | 72 | ||
| AK136553.1 | - | 5 - 6 | 957 | 72 | ||
| AK151346.1 | - | 5 - 6 | 956 | 72 | ||
| AK171676.1 | - | 5 - 6 | 938 | 72 | ||
| BC111882.1 | - | 4 - 5 | 895 | 40060245 | 72 | |
| BC112402.1 | - | 4 - 5 | 895 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifi44
Genomic Map