Assay Details
| Gene Symbol: Slc22a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133980.3 | NP_598741.2 | 3 - 4 | 679 | 101 | |
| GenBank mRNA | AF529220.1 | - | 3 - 4 | 672 | 101 | |
| BC021449.1 | - | 3 - 4 | 671 | 5044564 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977245 | BC021449.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977245 | AF529220.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977245 | NM_133980.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977245 | AI649135.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 968323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305297 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735127 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513632 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 733796 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106455 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79325 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106103 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153650 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977245 | BC021449.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977245 | AF529220.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977245 | NM_133980.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977245 | AI649135.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P154953 | BC021449.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P154953 | AF529220.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P154953 | AI649135.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P154953 | NM_133980.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871157 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106103 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02043999 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153650 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06946185 | - | - |
| Whole Mouse Genome G4122A | A_51_P154953 | BC021449.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154953 | AF529220.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154953 | AI649135.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154953 | NM_133980.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137358 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106103 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153650 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07799779 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08054943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07938221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07938225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07911955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08155489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07799779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07591866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08054943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871154 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10597617 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597616 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597615 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597618 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597621 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597614 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597613 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419129_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441723_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10597619 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10597612 | - | - |
| Mouse Genome 430 2.0 Array | 1419129_at | - | - |
| Mouse Genome 430 2.0 Array | 1419129_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 102570_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 102570_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900027215.mm.1 | NM_133980.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027215.mm.1 | NM_133980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027215.mm.1 | TC0900003236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900027213.mm.1 | NM_133980.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027213.mm.1 | NM_133980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027213.mm.1 | TC0900003236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900027221.mm.1 | NM_133980.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027221.mm.1 | NM_133980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027221.mm.1 | TC0900003236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900027216.mm.1 | NM_133980.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027216.mm.1 | NM_133980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027216.mm.1 | TC0900003236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900027219.mm.1 | NM_133980.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027219.mm.1 | NM_133980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027219.mm.1 | TC0900003236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900027217.mm.1 | NM_133980.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027217.mm.1 | NM_133980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027217.mm.1 | TC0900003236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900027220.mm.1 | NM_133980.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027220.mm.1 | NM_133980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027220.mm.1 | TC0900003236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900027222.mm.1 | NM_133980.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027222.mm.1 | NM_133980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027222.mm.1 | TC0900003236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900027214.mm.1 | NM_133980.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027214.mm.1 | NM_133980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027214.mm.1 | TC0900003236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900027218.mm.1 | NM_133980.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027218.mm.1 | NM_133980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027218.mm.1 | TC0900003236.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419129_PM_at | - | - |
| Mouse Expression Set 430 | 1419129_at | - | - |
| Mouse Expression Set 430 | 1441723_PM_at | - | - |
| Mouse Expression Set 430 | 1441723_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419129_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419129_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419129_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419129_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 523268 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1770563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001770563 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2682859 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2682859 | - | - |
| mouse-6 v1.1 (Illumina) | scl0102570.2_242-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1770563 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0102570.2_242-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1770563 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2682859 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3126190 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3050107 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2682859 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3126190 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3050107 | - | - |

Slc22a13
Genomic Map