Assay Details
| Gene Symbol: Apof | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133997.2 | NP_598758.1 | 1 - 2 | 775 | 69 | |
| GenBank mRNA | BC022795.1 | - | 1 - 2 | 74 | 5102555 | 69 |
| BC024769.1 | - | 1 - 2 | 62 | 5123598 | 69 | |
| BC025827.1 | - | 1 - 2 | 49 | 5101712 | 69 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC022795.1,BC024769.1,BC025827.1,NM_133997.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495825 | BC024769.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495825 | BC025827.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495825 | BC022795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495825 | NM_133997.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495825 | BC010815.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495825 | BC024769.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495825 | BC025827.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495825 | BC022795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495825 | NM_133997.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495825 | BC010815.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597638 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377307 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12442 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495825 | BC024769.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495825 | BC025827.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495825 | BC022795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495825 | NM_133997.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495825 | BC010815.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495825 | BC024769.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495825 | BC025827.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495825 | BC022795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495825 | NM_133997.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495825 | BC010815.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495825 | BC024769.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495825 | BC025827.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495825 | BC022795.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495825 | NM_133997.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495825 | BC010815.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495825 | BC024769.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495825 | BC025827.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495825 | BC022795.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495825 | NM_133997.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495825 | BC010815.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27540428 | - | - |
| Whole Mouse Genome G4122A | A_51_P495825 | BC024769.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495825 | BC025827.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495825 | BC022795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495825 | NM_133997.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495825 | BC010815.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495825 | BC024769.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495825 | BC025827.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495825 | BC022795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495825 | NM_133997.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495825 | BC010815.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135274 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08713343 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08713351 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08713347 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08713345 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08823581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27540474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08713346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08599771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08558679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08812722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08760122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08581609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08323527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08599769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27540445 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10367223 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367222 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418239_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10367221 | - | - |
| Mouse Genome 430 2.0 Array | 1418239_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418239_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103161_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103161_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000011659.mm.1 | NM_133997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011659.mm.1 | NM_133997.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011659.mm.1 | TC1000001595.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011664.mm.1 | NM_133997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011664.mm.1 | NM_133997.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011664.mm.1 | TC1000001595.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011663.mm.1 | NM_133997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011663.mm.1 | NM_133997.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011663.mm.1 | TC1000001595.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418239_at | - | - |
| Mouse Expression Set 430 | 1418239_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418239_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418239_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418239_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418239_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 825732 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5270707 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270707 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2644185 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2644185 | - | - |
| mouse-6 v1.1 (Illumina) | scl38289.2_239-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270707 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38289.2_239-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270707 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2644185 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2644185 | - | - |

Apof
Genomic Map