Assay Details
| Gene Symbol: Rdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134006.4 | NP_598767.1 | 3 - 4 | 747 | 100 | |
| GenBank mRNA | AF013288.1 | - | 3 - 4 | 618 | 100 | |
| AF033195.1 | - | 3 - 4 | 685 | 100 | ||
| AF033196.2 | - | 3 - 4 | 713 | 100 | ||
| AK135139.1 | - | 3 - 4 | 748 | 100 | ||
| BC021372.1 | - | 3 - 4 | 660 | 5040142 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P138289 | AF033196.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P138289 | BC021372.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P138289 | NM_134006.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P138289 | AF013288.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467714 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 97323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513877 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52714 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137852 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P138289 | AF033196.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P138289 | BC021372.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P138289 | NM_134006.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P138289 | AF013288.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P479618 | BC021372.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P479618 | AF013288.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543871 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137852 | - | - |
| Whole Mouse Genome G4122A | A_51_P479618 | BC021372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479618 | AF013288.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138289 | AF033196.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138289 | BC021372.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138289 | NM_134006.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138289 | AF013288.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137852 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08744353 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08744352 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08744348 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08744349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08744349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08518774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08954768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08954786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08895547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08744355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08954787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08895546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08895544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08404211 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373592 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373593 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373589 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373591 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373590 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418808_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373588 | - | - |
| Mouse Genome 430 2.0 Array | 1418808_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418808_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19682_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19682_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000023892.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023892.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023892.mm.1 | TC1000003227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023894.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023894.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023894.mm.1 | TC1000003227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000024056.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000024056.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000024056.mm.1 | TC1000003227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023893.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023893.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023893.mm.1 | TC1000003227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023902.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023902.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023902.mm.1 | TC1000003227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023896.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023896.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023896.mm.1 | TC1000003227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000024057.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000024057.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000024057.mm.1 | TC1000003227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023903.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023903.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023903.mm.1 | TC1000003227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023910.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023910.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023910.mm.1 | TC1000003227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023904.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023904.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023904.mm.1 | TC1000003227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023911.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023911.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023911.mm.1 | TC1000003227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023897.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023897.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023897.mm.1 | TC1000003227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023909.mm.1 | NM_134006.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023909.mm.1 | NM_134006.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023909.mm.1 | TC1000003227.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418808_PM_at | - | - |
| Mouse Expression Set 430 | 1418808_at | - | - |
| Murine Genome U74v2 Set | 101431_at | - | - |
| Murine Genome U74v2 Set | 101431_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418808_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418808_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418808_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418808_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 754399 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4610059 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106860497 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004610059 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221292 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221292 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253251 | - | - |
| mouse-6 v1.1 (Illumina) | scl37342.5.7_184-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4610059 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106860497 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37342.5.7_184-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4610059 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106860497 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221292 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253251 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221292 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37342.5.7_184-S | - | - |

Rdh5
Genomic Map