Assay Details
| Gene Symbol: Rcbtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170694.1 | NP_001164165.1 | 8 - 9 | 960 | 59 | |
| NM_134083.4 | NP_598844.3 | 8 - 9 | 955 | 59 | ||
| NR_033185.1 | 9 - 10 | 1340 | 59 | |||
| GenBank mRNA | AK031242.1 | - | 8 - 9 | 921 | 59 | |
| AK031248.1 | - | 8 - 9 | 926 | 59 | ||
| AK147784.1 | - | 9 - 10 | 961 | 59 | ||
| AK160881.1 | - | 9 - 10 | 1147 | 59 | ||
| BC003224.1 | - | 6 - 7 | 753 | 3482729 | 59 | |
| JN086338.1 | - | 7 - 8 | 876 | 59 | ||
| JN086339.1 | - | 8 - 9 | 1114 | 59 | ||
More Information
Set Membership: |
|

Rcbtb2
Genomic Map