Assay Details
| Gene Symbol: Slc38a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166456.1 | NP_001159928.1 | 8 - 9 | 956 | 63 | |
| NM_001166458.1 | NP_001159930.1 | 8 - 9 | 1163 | 63 | ||
| NM_134086.4 | NP_598847.2 | 8 - 9 | 1649 | 63 | ||
| GenBank mRNA | AF184240.2 | - | 7 - 8 | 741 | 63 | |
| AK029189.1 | - | 8 - 9 | 826 | 63 | ||
| AK034130.1 | - | 8 - 9 | 1651 | 63 | ||
| AK035098.1 | - | 7 - 8 | 668 | 63 | ||
| AK047459.1 | - | 8 - 9 | 1180 | 63 | ||
| AK049173.1 | - | 8 - 9 | 863 | 63 | ||
| AK049294.1 | - | 8 - 9 | 929 | 63 | ||
| AK050914.1 | - | 8 - 9 | 864 | 63 | ||
| AK081724.1 | - | 8 - 9 | 1165 | 63 | ||
| AK162612.1 | - | 8 - 9 | 1014 | 63 | ||
| AK163914.1 | - | 3 - 4 | 267 | 63 | ||
| AK165188.1 | - | 8 - 9 | 1216 | 63 | ||
| BC030378.1 | - | 8 - 9 | 865 | 5375319 | 63 | |
More Information
Set Membership: |
|

Slc38a1
Genomic Map