Assay Details
| Gene Symbol: Slc38a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166456.1 | NP_001159928.1 | 12 - 13 | 1306 | 66 | |
| NM_001166458.1 | NP_001159930.1 | 12 - 13 | 1513 | 66 | ||
| NM_134086.4 | NP_598847.2 | 12 - 13 | 1999 | 66 | ||
| GenBank mRNA | AF184240.2 | - | 11 - 12 | 1091 | 66 | |
| AK034130.1 | - | 12 - 13 | 2001 | 66 | ||
| AK035098.1 | - | 11 - 12 | 1018 | 66 | ||
| AK047459.1 | - | 12 - 13 | 1530 | 66 | ||
| AK049173.1 | - | 12 - 13 | 1214 | 66 | ||
| AK049294.1 | - | 12 - 13 | 1279 | 66 | ||
| AK081724.1 | - | 12 - 13 | 1515 | 66 | ||
| AK082683.1 | - | 12 - 13 | 1217 | 66 | ||
| AK162612.1 | - | 12 - 13 | 1364 | 66 | ||
| AK163914.1 | - | 7 - 8 | 617 | 66 | ||
| AK165188.1 | - | 12 - 13 | 1566 | 66 | ||
| BC030378.1 | - | 12 - 13 | 1215 | 5375319 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a1
Genomic Map