Assay Details
| Gene Symbol: Desi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134095.2 | NP_598856.1 | 1 - 2 | 263 | 121 | |
| GenBank mRNA | AK005706.1 | - | 1 - 2 | 254 | 121 | |
| AK005963.1 | - | 1 - 2 | 219 | 121 | ||
| AK016347.1 | - | 1 - 2 | 172 | 121 | ||
| AK131703.1 | - | 1 - 2 | 217 | 121 | ||
| AK146155.1 | - | 1 - 2 | 287 | 121 | ||
| AK170731.1 | - | 1 - 2 | 349 | 121 | ||
| BC056972.1 | - | 1 - 2 | 263 | 6811343 | 121 | |
| BC071205.1 | - | 1 - 2 | 155 | 6515068 | 121 | |
| BC131979.1 | - | 1 - 2 | 159 | 40130256 | 121 | |
| BC132642.1 | - | 1 - 2 | 159 | 40130919 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Desi1
Genomic Map