Assay Details
| Gene Symbol: Ildr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285788.1 | NP_001272717.1 | 4 - 5 | 674 | 74 | |
| NM_001285791.1 | NP_001272720.1 | 4 - 5 | 674 | 74 | ||
| NM_134109.2 | NP_598870.1 | 4 - 5 | 674 | 74 | ||
| GenBank mRNA | AK035504.1 | - | 4 - 5 | 603 | 74 | |
| AK050806.1 | - | 4 - 5 | 620 | 74 | ||
| AK136284.1 | - | 4 - 5 | 674 | 74 | ||
| BC010485.1 | - | 4 - 5 | 614 | 3483341 | 74 | |
| BC057644.1 | - | 4 - 5 | 671 | 4912789 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ildr1
Genomic Map