Assay Details
| Gene Symbol: Timd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161355.1 | NP_001154827.1 | 8 - 9 | 1090 | 111 | |
| NM_001161356.1 | NP_001154828.1 | 8 - 9 | 863 | 111 | ||
| NM_134249.5 | NP_599010.4 | 8 - 9 | 972 | 111 | ||
| GenBank mRNA | AF399828.1 | - | 8 - 9 | 824 | 111 | |
| AK145630.1 | - | 8 - 9 | 851 | 111 | ||
| AK163225.1 | - | 8 - 9 | 851 | 111 | ||
| AK166877.1 | - | 8 - 9 | 844 | 111 | ||
| BC025096.1 | - | 8 - 9 | 902 | 4158605 | 111 | |
| BC028829.1 | - | 8 - 9 | 934 | 5102129 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143030 | AK166877.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143030 | AK145630.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143030 | NM_134249.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143030 | NM_001161355.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143030 | NM_001161356.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053459 | AK166877.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053459 | BC028829.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053459 | AK145630.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053459 | AK163225.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053459 | NM_134249.5 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 283346 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506884 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 653830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937050 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884528 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693173 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191781 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200567 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915371 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737459 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111255 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343027 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404817 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630044 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 121359 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 122999 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109537 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118430 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118415 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148451 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120249 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053459 | AK166877.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053459 | BC028829.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053459 | AK145630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053459 | AK163225.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053459 | NM_134249.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143030 | AK166877.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143030 | AK145630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143030 | NM_134249.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143030 | NM_001161355.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143030 | NM_001161356.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27769343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27769348 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120249 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148451 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02316564 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07329610 | - | - |
| Whole Mouse Genome G4122A | A_52_P17556 | AK166877.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P17556 | BC028829.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P17556 | AK145630.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P17556 | AK163225.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P17556 | NM_134249.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500906 | AK166877.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500906 | BC025096.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500906 | BC028829.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500906 | AK145630.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500906 | AK163225.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104954 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148451 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27769173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09362809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09355079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09271093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09429358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27769305 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10385463 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385457 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385465 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385462 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385461 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385459 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385460 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385464 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418765_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418766_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10385455 | - | - |
| Mouse Genome 430 2.0 Array | 1418766_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1418765_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418766_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1418765_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 171284_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 171284_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100024034.mm.1 | NM_001161355.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024034.mm.1 | NM_001161355.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024034.mm.1 | NM_001161356.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024034.mm.1 | NM_001161356.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024034.mm.1 | NM_134249.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024034.mm.1 | NM_134249.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024034.mm.1 | TC1100002611.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100024033.mm.1 | NM_001161355.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024033.mm.1 | NM_001161355.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024033.mm.1 | NM_001161356.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024033.mm.1 | NM_001161356.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024033.mm.1 | NM_134249.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024033.mm.1 | NM_134249.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024033.mm.1 | TC1100002611.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100024035.mm.1 | NM_001161355.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024035.mm.1 | NM_001161355.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024035.mm.1 | NM_001161356.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024035.mm.1 | NM_001161356.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024035.mm.1 | NM_134249.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024035.mm.1 | NM_134249.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024035.mm.1 | TC1100002611.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100024031.mm.1 | NM_001161355.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024031.mm.1 | NM_001161355.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024031.mm.1 | NM_001161356.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024031.mm.1 | NM_001161356.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024031.mm.1 | NM_134249.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024031.mm.1 | NM_134249.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024031.mm.1 | TC1100002611.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418766_PM_s_at | - | - |
| Mouse Expression Set 430 | 1418766_s_at | - | - |
| Mouse Expression Set 430 | 1418765_PM_at | - | - |
| Mouse Expression Set 430 | 1418765_at | - | - |
| Murine Genome U74v2 Set | 97335_at | - | - |
| Murine Genome U74v2 Set | 103794_i_at | - | - |
| Murine Genome U74v2 Set | 103794_PM_i_at | - | - |
| Murine Genome U74v2 Set | 103795_f_at | - | - |
| Murine Genome U74v2 Set | 103795_PM_f_at | - | - |
| Murine Genome U74v2 Set | 97335_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418766_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418765_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418765_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418766_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418766_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418765_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418765_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418766_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 642013 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5690494 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005690494 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2773415 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2773415 | - | - |
| mouse-6 v1.1 (Illumina) | scl0171284.1_49-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5690494 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0171284.1_49-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690494 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2940135 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2940135 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2773415 | - | - |

Timd2
Genomic Map