Assay Details
| Gene Symbol: Slc12a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083902.1 | NP_001077371.1 | 3 - 4 | 422 | 77 | |
| NM_134251.2 | NP_599012.2 | 3 - 4 | 391 | 77 | ||
| GenBank mRNA | AF319951.1 | - | 3 - 4 | 385 | 77 | |
| BC030926.1 | - | 3 - 4 | 422 | 4910321 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a8
Genomic Map