Assay Details
| Gene Symbol: Ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138589.2 | NP_613055.2 | 3 - 4 | 829 | 109 | |
| GenBank mRNA | AK011862.1 | - | 3 - 4 | 585 | 109 | |
| AK017851.1 | - | 3 - 4 | 606 | 108 | ||
| AK033428.1 | - | 2 - 3 | 1608 | 109 | ||
| AK077395.1 | - | 3 - 4 | 852 | 109 | ||
| AK140891.1 | - | 3 - 4 | 588 | 109 | ||
| BC011313.1 | - | 3 - 4 | 832 | 3499738 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubfd1
Genomic Map