Assay Details
| Gene Symbol: Fam50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138607.3 | NP_613073.2 | 11 - 12 | 1083 | 77 | |
| GenBank mRNA | AK008402.1 | - | 11 - 12 | 1084 | 77 | |
| BC125297.1 | - | 10 - 11 | 949 | 40129812 | 77 | |
| Y18505.1 | - | 11 - 12 | 1048 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam50a
Genomic Map