Assay Details
| Gene Symbol: Luc7l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170848.1 | NP_001164319.1 | 4 - 5 | 772 | 86 | |
| NM_001170849.1 | NP_001164320.1 | 4 - 5 | 772 | 86 | ||
| NM_138680.2 | NP_619621.2 | 4 - 5 | 772 | 86 | ||
| GenBank mRNA | AF318301.1 | - | 4 - 5 | 697 | 86 | |
| AK134191.1 | - | 4 - 5 | 686 | 86 | ||
| AK147889.1 | - | 7 - 8 | 943 | 86 | ||
| AK153527.1 | - | 8 - 9 | 1029 | 86 | ||
| BC020058.1 | - | 5 - 6 | 497 | 4009362 | 86 | |
| BC040793.1 | - | 3 - 4 | 264 | 86 | ||
| BC054449.1 | - | 3 - 4 | 715 | 86 | ||
| BC056354.1 | - | 4 - 5 | 770 | 6811460 | 86 | |
| BC056383.1 | - | 4 - 5 | 751 | 6853955 | 86 | |
| BC056970.1 | - | 4 - 5 | 770 | 6811460 | 86 | |
More Information
Set Membership: |
|

Luc7l2
Genomic Map