| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200010656.mm.1 |
NM_001004455.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010656.mm.1 |
NM_001004455.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010656.mm.1 |
NM_001161807.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010656.mm.1 |
NM_001161807.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010656.mm.1 |
NM_138686.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010656.mm.1 |
NM_138686.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010656.mm.1 |
TC1200001536.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200010658.mm.1 |
NM_138686.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010658.mm.1 |
NM_138686.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010658.mm.1 |
TC1200001536.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200010657.mm.1 |
NM_001161807.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010657.mm.1 |
NM_001161807.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010657.mm.1 |
NM_138686.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010657.mm.1 |
NM_138686.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010657.mm.1 |
TC1200001536.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200010661.mm.1 |
NM_001004455.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010661.mm.1 |
NM_001004455.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010661.mm.1 |
NM_001161807.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010661.mm.1 |
NM_001161807.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010661.mm.1 |
NM_138686.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010661.mm.1 |
NM_138686.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010661.mm.1 |
TC1200001536.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200010654.mm.1 |
NM_001004455.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010654.mm.1 |
NM_001004455.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010654.mm.1 |
NM_001161807.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010654.mm.1 |
NM_001161807.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010654.mm.1 |
NM_138686.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010654.mm.1 |
NM_138686.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010654.mm.1 |
TC1200001536.mm.1 |
MTA TC |