Assay Details
| Gene Symbol: Tmem38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144534.1 | NP_653117.1 | 2 - 3 | 282 | 86 | |
| GenBank mRNA | AB261158.1 | - | 2 - 3 | 281 | 86 | |
| AK157969.1 | - | 2 - 3 | 342 | 86 | ||
| AK165818.1 | - | 3 - 4 | 342 | 86 | ||
| AK186226.1 | - | 1 - 2 | 140 | 86 | ||
| AK210187.1 | - | 1 - 2 | 140 | 86 | ||
| AK215082.1 | - | 1 - 2 | 140 | 86 | ||
| BC014728.1 | - | 2 - 3 | 295 | 4504294 | 86 | |
| BC024788.1 | - | 2 - 3 | 282 | 5360106 | 86 | |
| EF988666.1 | - | 2 - 3 | 316 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem38a
Genomic Map