Assay Details
| Gene Symbol: Tmem38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144534.1 | NP_653117.1 | 4 - 5 | 547 | 89 | |
| GenBank mRNA | AB261158.1 | - | 4 - 5 | 546 | 89 | |
| AK157969.1 | - | 4 - 5 | 607 | 89 | ||
| AK165818.1 | - | 5 - 6 | 607 | 89 | ||
| BC014728.1 | - | 4 - 5 | 560 | 4504294 | 89 | |
| BC024788.1 | - | 4 - 5 | 547 | 5360106 | 89 | |
| EF988666.1 | - | 4 - 5 | 581 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem38a
Genomic Map