Assay Details
| Gene Symbol: Appl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145220.2 | NP_660255.1 | 1 - 2 | 139 | 90 | |
| GenBank mRNA | AK154525.1 | - | 1 - 2 | 163 | 90 | |
| AK160075.1 | - | 1 - 2 | 118 | 90 | ||
| AK160105.1 | - | 1 - 2 | 115 | 90 | ||
| AK161003.1 | - | 1 - 2 | 146 | 90 | ||
| AK168403.1 | - | 1 - 2 | 100 | 90 | ||
| AK168748.1 | - | 1 - 2 | 95 | 90 | ||
| AK171512.1 | - | 1 - 2 | 95 | 90 | ||
| AY113706.1 | - | 1 - 2 | 141 | 90 | ||
| BC048906.1 | - | 1 - 2 | 77 | 6475576 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Appl2
Genomic Map