Assay Details
| Gene Symbol: Aars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146217.4 | NP_666329.2 | 14 - 15 | 2434 | 95 | |
| GenBank mRNA | AK085725.1 | - | 14 - 15 | 2375 | 95 | |
| AK150227.1 | - | 14 - 15 | 2362 | 95 | ||
| AK196199.1 | - | 1 - 2 | 95 | 95 | ||
| AK214611.1 | - | 1 - 2 | 60 | 95 | ||
| AY223875.1 | - | 14 - 15 | 2374 | 95 | ||
| BC013261.1 | - | 11 - 12 | 1659 | 95 | ||
| BC026611.1 | - | 3 - 4 | 463 | 95 | ||
| BC033273.1 | - | 2309 | 4196789 | 95 | ||
| BC058620.1 | - | 4 - 5 | 608 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aars
Genomic Map