Assay Details
| Gene Symbol: Unc5cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152823.4 | NP_690036.4 | 4 - 5 | 1077 | 63 | |
| GenBank mRNA | AK039924.1 | - | 1 - 2 | 75 | 63 | |
| AK089034.1 | - | 1 - 2 | 50 | 63 | ||
| AK144925.1 | - | 4 - 5 | 1078 | 63 | ||
| AY510108.1 | - | 3 - 4 | 944 | 63 | ||
| BC022701.1 | - | 1 - 2 | 99 | 63 | ||
| BC095959.1 | - | 4 - 5 | 1055 | 5349225 | 63 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK039924.1,AK089034.1,BC022701.1 transcripts.
Set Membership: |
|

Unc5cl
Genomic Map