Assay Details
| Gene Symbol: Arnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037737.2 | NP_001032826.1 | 6 - 7 | 630 | 86 | |
| NM_009709.4 | NP_033839.2 | 5 - 6 | 585 | 86 | ||
| GenBank mRNA | AK028546.1 | - | 5 - 6 | 572 | 86 | |
| AK040475.1 | - | 6 - 7 | 632 | 86 | ||
| AK141362.1 | - | 6 - 7 | 501 | 86 | ||
| AK145415.1 | - | 5 - 6 | 540 | 86 | ||
| AK152473.1 | - | 6 - 7 | 612 | 86 | ||
| AK153355.1 | - | 6 - 7 | 612 | 86 | ||
| AK220458.1 | - | 2 - 3 | 761 | 86 | ||
| BC012870.1 | - | 6 - 7 | 592 | 3964932 | 86 | |
| U10325.1 | - | 6 - 7 | 494 | 86 | ||
| U14333.1 | - | 5 - 6 | 423 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arnt
Genomic Map