Assay Details
| Gene Symbol: Arnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037737.2 | NP_001032826.1 | 8 - 9 | 951 | 109 | |
| NM_009709.4 | NP_033839.2 | 7 - 8 | 906 | 109 | ||
| GenBank mRNA | AK028546.1 | - | 7 - 8 | 893 | 109 | |
| AK040475.1 | - | 8 - 9 | 953 | 109 | ||
| AK141362.1 | - | 8 - 9 | 822 | 109 | ||
| AK145415.1 | - | 7 - 8 | 861 | 109 | ||
| AK152473.1 | - | 8 - 9 | 933 | 109 | ||
| AK153355.1 | - | 8 - 9 | 933 | 109 | ||
| AK220458.1 | - | 4 - 5 | 1082 | 109 | ||
| BC012870.1 | - | 8 - 9 | 913 | 3964932 | 109 | |
| U10325.1 | - | 8 - 9 | 815 | 109 | ||
| U14333.1 | - | 7 - 8 | 744 | 109 | ||
More Information
Set Membership: |
|

Arnt
Genomic Map