Assay Details
| Gene Symbol: Arnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037737.2 | NP_001032826.1 | 17 - 18 | 1845 | 73 | |
| NM_009709.4 | NP_033839.2 | 16 - 17 | 1800 | 73 | ||
| GenBank mRNA | AK028546.1 | - | 16 - 17 | 1787 | 73 | |
| AK040475.1 | - | 17 - 18 | 1847 | 73 | ||
| AK145415.1 | - | 16 - 17 | 1755 | 73 | ||
| AK152473.1 | - | 17 - 18 | 1827 | 73 | ||
| AK153355.1 | - | 17 - 18 | 1827 | 73 | ||
| AK220458.1 | - | 13 - 14 | 1976 | 73 | ||
| BC012870.1 | - | 17 - 18 | 1807 | 3964932 | 73 | |
| U10325.1 | - | 17 - 18 | 1709 | 73 | ||
| U14333.1 | - | 16 - 17 | 1638 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arnt
Genomic Map