Assay Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011790.4 | NP_035920.1 | 14 - 15 | 1626 | 123 | |
| GenBank mRNA | AF124664.1 | - | 13 - 14 | 1453 | 123 | |
| AJ130975.1 | - | 13 - 14 | 1485 | 123 | ||
| AK011094.1 | - | 14 - 15 | 1633 | 123 | ||
| AK045012.1 | - | 1609 | 123 | |||
| AK149936.1 | - | 14 - 15 | 1606 | 123 | ||
| AK167104.1 | - | 14 - 15 | 1583 | 123 | ||
| AK168055.1 | - | 14 - 15 | 1629 | 123 | ||
| BC051998.1 | - | 14 - 15 | 1626 | 6400287 | 123 | |
| BC052422.1 | - | 14 - 15 | 1626 | 6400287 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arih2
Genomic Map