Assay Details
| Gene Symbol: Isoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025478.3 | NP_079754.2 | 2 - 3 | 435 | 126 | |
| GenBank mRNA | AK011674.1 | - | 2 - 3 | 302 | 126 | |
| AK134794.1 | - | 2 - 3 | 435 | 126 | ||
| AK152763.1 | - | 2 - 3 | 432 | 126 | ||
| AK153257.1 | - | 2 - 3 | 432 | 126 | ||
| AK165542.1 | - | 2 - 3 | 454 | 126 | ||
| AY035213.1 | - | 2 - 3 | 435 | 126 | ||
| BC016576.1 | - | 2 - 3 | 442 | 2609919 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Isoc1
Genomic Map