Assay Details
| Gene Symbol: Dhrs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025522.5 | NP_079798.2 | 1 - 2 | 197 | 65 | |
| GenBank mRNA | AK009385.1 | - | 1 - 2 | 176 | 65 | |
| AK014100.1 | - | 1 - 2 | 152 | 65 | ||
| BC016189.1 | - | 1 - 2 | 163 | 4482814 | 65 | |
| CX223953.1 | - | 1 - 2 | 197 | 65 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK014100.1,BC016189.1 transcripts.
Set Membership: |
|

Dhrs7
Genomic Map