Assay Details
| Gene Symbol: Cmss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025599.3 | NP_079875.2 | 6 - 7 | 649 | 138 | |
| GenBank mRNA | AK012168.1 | - | 6 - 7 | 630 | 138 | |
| AK019979.1 | - | 6 - 7 | 625 | 138 | ||
| BC038891.1 | - | 6 - 7 | 536 | 3371329 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cmss1
Genomic Map