Assay Details
| Gene Symbol: Cmss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025599.3 | NP_079875.2 | 7 - 8 | 718 | 97 | |
| GenBank mRNA | AK012168.1 | - | 7 - 8 | 699 | 97 | |
| AK019979.1 | - | 7 - 8 | 694 | 97 | ||
| BC038891.1 | - | 7 - 8 | 605 | 3371329 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cmss1
Genomic Map