Assay Details
| Gene Symbol: Cmss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025599.3 | NP_079875.2 | 8 - 9 | 806 | 111 | |
| GenBank mRNA | AK012168.1 | - | 8 - 9 | 787 | 111 | |
| AK019979.1 | - | 8 - 9 | 782 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cmss1
Genomic Map