Assay Details
| Gene Symbol: Pacrgl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025755.3 | NP_080031.1 | 5 - 6 | 483 | 98 | |
| GenBank mRNA | AK014276.1 | - | 5 - 6 | 496 | 98 | |
| AK016966.1 | - | 5 - 6 | 532 | 98 | ||
| AK018101.1 | - | 5 - 6 | 493 | 97 | ||
| AK032498.1 | - | 4 - 5 | 2231 | 98 | ||
| BC018481.1 | - | 5 - 6 | 483 | 3482169 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pacrgl
Genomic Map